Perfluoroheptanoic acid ≥98.0% (by titrimetric analysis), High Grade per cromatografia, per LC-MS
Fornitore: TCI
|
Pericolo
|
Formula:
C₇HF₁₃O₂ Punto di ebollizione: 177°C |
MDL Number:
MFCD00039604 CAS Number: 375-85-9 EINECS: 206-798-9 UN: 3261 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (Neutralization titration) | min. 98.0 % |
Absorbance (230nm) | max. 0.20 |
Absorbance (254nm) | max. 0.01 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...