Stampa…

4-Pyridineboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Pyridinylboronic acid, Pyridine-4-boronic acid, Pyridin-4-ylboronic acid

P1594-1G P1594-5G
TCIAP1594-1GEA 18 EUR
TCIAP1594-1G TCIAP1594-5G
4-Pyridineboronic acid (contains varying amounts of Anhydride)
4-Pyridineboronic acid
Formula: C₅H₆BNO₂
Peso molecolare: 122,92 g/mol (anhy)
MDL Number: MFCD01074545
CAS Number: 1692-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8fd8712a8c1db8667d767645fb6cff10cfb68db&vid=c8fd8712a8c1db8667d767645fb6cff10cfb68db&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 96.0 to 117.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR