3-Pyridineboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Pyridinylboronic acid, Pyridin-3-ylboronic acid, Pyridine-3-boronic acid
P1673-1G
P1673-25G
P1673-5G
TCIAP1673-1GEA
35
EUR
TCIAP1673-1G
TCIAP1673-25G
TCIAP1673-5G
3-Pyridineboronic acid (contains varying amounts of Anhydride)
3-Pyridineboronic acid
Formula:
C₅H₆BNO₂ Peso molecolare: 122,92 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00674177 CAS Number: 1692-25-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 117.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...