2-Bromo-5-pyridineboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Bromo-3-pyridinylboronic acid, 2-Bromopyridine-5-boronic acid, (6-Bromopyridin-3-yl)boronic acid, 2-Bromo-5-pyridinylboronic acid, 6-Bromopyridine-3-boronic acid
B3307-1G
B3307-5G
TCIAB3307-1GEA
57
EUR
TCIAB3307-1G
TCIAB3307-5G
2-Bromo-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Bromo-5-pyridineboronic acid
Formula:
C₅H₅BBrNO₂ Peso molecolare: 201,81 g/mol |
MDL Number:
MFCD03411558 CAS Number: 223463-14-7 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | 96.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...