Stampa…

1-Ethyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis) for molten salt

Fornitore: TCI
Attenzione

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride

E0490-25G E0490-250G E0490-5G
TCIAE0490-25GEA 53 EUR
TCIAE0490-25G TCIAE0490-250G TCIAE0490-5G
1-Ethyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis) for molten salt
1-Ethyl-3-methylimidazolium chloride
Formula: C₆H₁₁ClN₂
Peso molecolare: 146.62 g/mol
Punto di fusione: 84°C
Flash Pt: 186
MDL Number: MFCD00074843
CAS Number: 65039-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c46c9dfe7cfdd94c57aea649b735b188ea72a2a3&vid=cc0755386a6dec74115de8b54a3910bb02035675&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR