Stampa…

Tricarballylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

P0490-25G P0490-100G
TCIAP0490-25GEA 92 EUR
TCIAP0490-25G TCIAP0490-100G
Tricarballylic acid ≥98.0% (by GC, titration analysis)
Tricarballylic acid
Formula: C₆H₈O₆
Punto di ebollizione: 266 °C (1013 hPa)
Punto di fusione: 160 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002723
CAS Number: 99-14-9
EINECS: 202-733-3
Merck Index: 14,09622
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fdd3c56040008cb679d8aff9115581d5c13ed96&vid=4fdd3c56040008cb679d8aff9115581d5c13ed96&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 163.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR