Potassium indole-3-acetate ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
Synonyms:
K-IAA, 3-Indoleacetic acid potassium salt, Heteroauxine, Indol-3-ylacetic acid potassium salt, Indole-3-acetic acid potassium salt, Potassium 3-Indoleacetate
Formula:
C₁₀H₈KNO₂ Peso molecolare: 213.28 g/mol |
MDL Number:
MFCD00066213 CAS Number: 2338-19-4 EINECS: 219-049-6 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...