Stampa…

Ruthenium(III) acetylacetonate

Fornitore: TCI
Attenzione

Synonyms: Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)

T2183-1G T2183-5G
TCIAT2183-1GEA 92 EUR
TCIAT2183-1G TCIAT2183-5G
Ruthenium(III) acetylacetonate
Ruthenium(III) acetylacetonate
Formula: C₁₅H₂₁O₆Ru
Peso molecolare: 398.4 g/mol
MDL Number: MFCD00000030
CAS Number: 14284-93-6
EINECS: 238-193-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0772734611c16f3167c39a10befe5a82bd29beab&vid=0772734611c16f3167c39a10befe5a82bd29beab&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Dark red powder to crystal
Elemental analysis (Carbon) 43.00 to 47.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR