Stampa…

Iron(III) acetylacetonate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

I0079-25G I0079-500G I0079-100G
TCIAI0079-25GEA 16 EUR
TCIAI0079-25G TCIAI0079-500G TCIAI0079-100G
Iron(III) acetylacetonate ≥98.0% (by titrimetric analysis)
Iron(III) acetylacetonate
Formula: C₁₅H₂₁FeO₆
Peso molecolare: 353,18 g/mol
Punto di ebollizione: 100 °C (9,7513 -rr)
Punto di fusione: 180…183 °C
Densità: 5,24 g/cm³ (20 °C)
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d417943775d10556d56627005a2062df8d24c5d&vid=4d417943775d10556d56627005a2062df8d24c5d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Red to Dark red to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR