Stampa…

Chromium(III) acetylacetonate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tris(2,4-pentanedionato-O,O')chromium (III), Chromium (III) 2,4-pentanedionate, Chromium (III) tris(2,4-pentanedionato-O,O')

C0336-25G C0336-100G
TCIAC0336-25GEA 17 EUR
TCIAC0336-25G TCIAC0336-100G
Chromium(III) acetylacetonate ≥98.0% (by titrimetric analysis)
Chromium(III) acetylacetonate
Formula: C₁₅H₂₁CrO₆
Peso molecolare: 349,32 g/mol
Punto di ebollizione: 340 °C (1013 hPa)
Punto di fusione: 215,85 °C
Densità: 1,266 g/cm³ (20 °C)
MDL Number: MFCD00000015
CAS Number: 21679-31-2
EINECS: 244-526-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b4950f94836339dc93163087b0986591bc07353&vid=9b4950f94836339dc93163087b0986591bc07353&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Gray to Dark purple to Black powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR