Stampa…

Dansyl amide ≥98.0% (by titrimetric analysis) for fluorometry

Fornitore: TCI EUROPE

Synonyms: Dansylamide, 5-(Dimethylamino)-1-naphthalenesulphonamide, 5-(Dimethylamino)naphthalene-1-sulphonamide

D1507-1G D1507-5G
TCIAD1507-1GEA 45 EUR
TCIAD1507-1G TCIAD1507-5G
Dansyl amide ≥98.0% (by titrimetric analysis) for fluorometry
Dansyl amide
Formula: C₁₂H₁₄N₂O₂S
Peso molecolare: 250,32 g/mol
Punto di ebollizione: 405 °C (1013 hPa)
Punto di fusione: 215 °C
MDL Number: MFCD00004000
CAS Number: 1431-39-6
EINECS: 215-854-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d0f14a1afd2c6318ad281d81c143d4f31134f52&vid=6a1f4dee6de86eb8abaec05b2c12475c4cc0c5ae&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 218.0 to 222.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR