Tetraetilammonio idrossido 35% in water
Fornitore: TCI
|
Pericolo
|
Synonyms:
Tetrylammonium hydroxide
Formula:
C₈H₂₁NO |
MDL Number:
MFCD00009024 CAS Number: 77-98-5 EINECS: 201-073-3 UN: 3267 ADR: 8,II Merck Index: 14,09201 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Neutralization titration) | 34.0 to 36.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...