Streptomicina solfato ≥95.0% (by total nitrogen basis) for protein research
Fornitore: TCI
|
Attenzione
|
S0834-25G
S0834-5G
TCIAS0834-25GEA
28
EUR
TCIAS0834-25G
TCIAS0834-5G
Streptomicina solfato ≥95.0% (by total nitrogen basis) for protein research
Streptomycin sulphate
Formula:
C₂₁H₃₉N₇O₁₂·1.5H₂SO₄ Punto di fusione: 250 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00037023 CAS Number: 3810-74-0 EINECS: 223-286-0 ADR: 6.1,III Merck Index: 14,08826 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % (calcd.on dried substance) |
Specific rotation [a]20/D | -79.0 to -88.0 deg (C=1, H2O) (calcd.on dried substance) |
Drying loss | max. 5.0 % |
Nucleic acid precipitation test | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...