Stampa…

L(-)-Glucosio ≥98.0% (by GC)

Fornitore: TCI

Synonyms: (2S,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal

G0226-5G G0226-1G
TCIAG0226-5GEA 311 EUR
TCIAG0226-5G TCIAG0226-1G
L(-)-Glucosio ≥98.0% (by GC)
L(-)-Glucosio
Formula: C₆H₁₂O₆
Peso molecolare: 180.16 g/mol
Punto di ebollizione: 376
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00148913
CAS Number: 921-60-8
EINECS: 213-068-3
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d65657a21d35dfa849425e1d1c8b44688e6c28c3&vid=d65657a21d35dfa849425e1d1c8b44688e6c28c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 % (total of isomers)
Specific rotation [a]20/D -50.0 to -54.0 deg (C=5, H2O:NH3 (2+3) = 99.2:0.8)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR