Stampa…

4-Nitrophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Nitrobenzeneboronic acid, (4-Nitrophenyl)boronic acid, Dihydroxy(4-nitrophenyl)borane, (4-Nitrophenyl)metaboric acid

N0812-1G N0812-5G
TCIAN0812-1GEA 27 EUR
TCIAN0812-1G TCIAN0812-5G
4-Nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
Peso molecolare: 166,93 g/mol
Punto di fusione: 305°C
MDL Number: MFCD00161360
CAS Number: 24067-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65a3b9945e4d14c85daa35ee3689b66a269b9ef5&vid=65a3b9945e4d14c85daa35ee3689b66a269b9ef5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Green to Brown powder to crystal
Purity (Neutralization titration) 97.0 to 112.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR