Stampa…

4-Chlorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

C1473-25G C1473-5G
TCIAC1473-25GEA 74 EUR
TCIAC1473-25G TCIAC1473-5G
4-Chlorophenylboronic acid (contains varying amounts of Anhydride)
4-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
Punto di fusione: 268…272 °C
Densità: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=094ff69af30367ddd19a53021e19d5901e99d81f&vid=094ff69af30367ddd19a53021e19d5901e99d81f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR