Stampa…

3-Methoxyphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (3-Methoxyphenyl)metaboric acid, Dihydroxy(3-methoxyphenyl)borane, 3-Methoxybenzeneboronic acid

M1322-1G M1322-25G M1322-5G
TCIAM1322-1GEA 30 EUR
TCIAM1322-1G TCIAM1322-25G TCIAM1322-5G
3-Methoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Methoxyphenylboronic acid
Formula: C₇H₉BO₃
Peso molecolare: 151,96 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00161359
CAS Number: 10365-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f143561effd4c57e49e912518fd6e4ca1228c6d5&vid=f143561effd4c57e49e912518fd6e4ca1228c6d5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR