Stampa…

1-Phenyl-1-octadecanone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Octadecanophenone, n-Octadecanophenone

O0185-25G O0185-5G
TCIAO0185-25GEA 177 EUR
TCIAO0185-25G TCIAO0185-5G
1-Phenyl-1-octadecanone ≥98.0% (by GC)
1-Phenyl-1-octadecanone
Formula: C₆H₅C(O)(CH₂)₁₆CH₃
Peso molecolare: 344,58 g/mol
Punto di ebollizione: 243°C
MDL Number: MFCD00010700
CAS Number: 6786-36-3
EINECS: 229-850-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a88aa57bfca5e1816877b435b5645a94c8bd7ae9&vid=a88aa57bfca5e1816877b435b5645a94c8bd7ae9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 67.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR