Stampa…
2,3-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
Attenzione
D3434-25G D3434-5G
TCIAD3434-25GEA 137 EUR
TCIAD3434-25G TCIAD3434-5G
2,3-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
2,3-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
Peso molecolare: 190,82 g/mol
MDL Number: MFCD01075703
CAS Number: 151169-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1762e8a8b1c0cf5b533ce3ec29993a246f97c6a5&vid=1762e8a8b1c0cf5b533ce3ec29993a246f97c6a5&prid=web&clid=SAPDAM
Visualizza i codici
Purity (Neutralization titration) 97.0 to 111.0 %