1-Butanesulphonic acid sodium salt ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butansulphonate, Butane-1-sulphonic acid sodium salt
Formula:
C₄H₉NaO₃S Punto di fusione: > 300 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007540 CAS Number: 2386-54-1 EINECS: 219-201-1 |
|
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...