(±)-1-Feniletanolo ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
(±)-sec-Phenethyl alcohol, Styrene alcohol, (±)-α-Methylbenzyl alcohol, (±)-1-Phenylethyl alcohol, 1-Phenylethanol, Methyl phenyl carbinol, α-Methylbenzyl alcohol, Styrallyl alcohol
Formula:
C₆H₅CH(OH)CH₃ Punto di ebollizione: 205°C Punto di fusione: 20 °C Densità: 1,015 g/cm³ (20 °C) Flash Pt: 94°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004508 CAS Number: 98-85-1 EINECS: 202-707-1 UN: 2937 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...