Tiourea ≥99.0% (by titrimetric analysis) for biochemical research
Fornitore: TCI
|
Attenzione
|
Formula:
NH₂CSNH₂ Punto di fusione: 177°C Densità: 1,435 g/cm³ (20 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 EINECS: 200-543-5 UN: 2811 ADR: 6.1,III Merck Index: 14,09367 |
|
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Iodometric back titration) | min. 99.0 % |
Suitability for protein-analysis | to pass test |
Protein denaturation test | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...