2-Acetylbenzo[b]thiophene ≥98.0% (by GC)
Fornitore: TCI
|
|
Formula:
C₁₀H₈OS |
MDL Number:
MFCD00090217 CAS Number: 22720-75-8 EINECS: 245-177-7 UN: 3077 ADR: 9,III |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 87.0 to 90.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...