1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione
T1583-25G
T1583-10G
T1583-5G
TCIAT1583-25GEA
110
EUR
TCIAT1583-25G
TCIAT1583-10G
TCIAT1583-5G
1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥98.0% (by GC)
1-(2-Naphthoyl)-3,3,3-trifluoroacetone
Formula:
C₁₄H₉F₃O₂ Peso molecolare: 266,22 g/mol |
MDL Number:
MFCD00054501 CAS Number: 893-33-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 73.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...