Stampa…

2-Amino-1-naphthol hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE
Attenzione

A0365-1G A0365-5G
TCIAA0365-1GEA 51 EUR
TCIAA0365-1G TCIAA0365-5G
2-Amino-1-naphthol hydrochloride ≥98.0% (by total nitrogen and titration analysis)
2-Amino-1-naphthol hydrochloride
Formula: C₁₀H₁₀ClNO
MDL Number: MFCD00050530
CAS Number: 41772-23-0
EINECS: 255-548-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=346dac903914dc1b7a720f1bd6c9ad682564d703&vid=346dac903914dc1b7a720f1bd6c9ad682564d703&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR