Stampa…
3-Methoxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Attenzione
Synonyms: 3-Methoxynaphthalene-2-carboxylic acid
M1461-1G M1461-5G
TCIAM1461-1GEA 64 EUR
TCIAM1461-1G TCIAM1461-5G
3-Methoxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
3-Methoxy-2-naphthoic acid
Formula: C₁₂H₁₀O₃
Peso molecolare: 202.2060 g/mol
MDL Number: MFCD00094168
CAS Number: 883-62-5
EINECS: 212-932-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbdad79b2e2a506b72509ccefda69d90992f7356&vid=bbdad79b2e2a506b72509ccefda69d90992f7356&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C