Stampa…

Methyl-6-amino-2-naphthoate ≥98.0% (by GC, titration analysis)

Fornitore: TCI

Synonyms: 6-Amino-2-naphthoic acid methyll ester, Methyl 6-amino-2-naphthoate

A1643-1G A1643-5G
TCIAA1643-1GEA 94 EUR
TCIAA1643-1G TCIAA1643-5G
Methyl-6-amino-2-naphthoate ≥98.0% (by GC, titration analysis)
Methyl-6-amino-2-naphthoate
Formula: C₁₂H₁₁NO₂
MDL Number: MFCD06797040
CAS Number: 5159-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c26797e90c72dfc8556e8d7a169b27b305526a53&vid=c26797e90c72dfc8556e8d7a169b27b305526a53&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 162.0 to 167.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR