N-Methyldicyclohexylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₃H₂₅N Punto di ebollizione: 150°C |
MDL Number:
MFCD00014289 CAS Number: 7560-83-0 EINECS: 231-453-4 UN: 2735 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...