Stampa…

Methyl-2-amino-5-iodobenzoate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-5-iodobenzoic acid methyll ester, Methyl 2-amino-5-iodobenzoate, Methyl 5-Iodoanthranilate, Methyl 2-Amino-5-iodobenzoate, 5-Iodoanthranilic acid methyll ester

I0516-25G I0516-5G
TCIAI0516-25GEA 168 EUR
TCIAI0516-25G TCIAI0516-5G
Methyl-2-amino-5-iodobenzoate ≥98.0% (by GC)
Methyl-2-amino-5-iodobenzoate
Formula: C₈H₈INO₂
Peso molecolare: 277,0591 g/mol
MDL Number: MFCD00798574
CAS Number: 77317-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38d82ef012e8392dc285d70679c2793d3d49be9c&vid=38d82ef012e8392dc285d70679c2793d3d49be9c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR