4-(4-Chlorophenoxy)aniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
4-Amino-4'-chlorodiphenyl ether
C1069-25G
C1069-5G
TCIAC1069-25GEA
137
EUR
TCIAC1069-25G
TCIAC1069-5G
4-(4-Chlorophenoxy)aniline ≥98.0% (by GC, titration analysis)
4-(4-Chlorophenoxy)aniline
Formula:
C₁₂H₁₀ClNO Peso molecolare: 219,67 g/mol Punto di ebollizione: 205°C |
MDL Number:
MFCD00043925 CAS Number: 101-79-1 EINECS: 202-976-5 |
Risultati del test di specificazione
Appearance | White to Light gray to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...