Stampa…

Methyl-2-amino-5-bromobenzoate ≥99.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 2-amino-5-bromobenzoate

B1988-25G B1988-5G
TCIAB1988-25GEA 135 EUR
TCIAB1988-25G TCIAB1988-5G
Methyl-2-amino-5-bromobenzoate ≥99.0% (by GC)
Methyl-2-amino-5-bromobenzoate
Formula: C₈H₈BrNO₂
Peso molecolare: 230,06 g/mol
MDL Number: MFCD00144761
CAS Number: 52727-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f50289a357e98328a523483d73dc9780ebe72456&vid=f50289a357e98328a523483d73dc9780ebe72456&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 70.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR