Stampa…

Ethyl-3-amino-4,4,4-trifluorocrotonate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 3-amino-4,4,4-trifluorobut-2-enoate

A1475-25G A1475-5G
TCIAA1475-25GEA 67 EUR
TCIAA1475-25G TCIAA1475-5G
Ethyl-3-amino-4,4,4-trifluorocrotonate ≥97.0% (by GC)
Ethyl-3-amino-4,4,4-trifluorocrotonate
Formula: C₆H₈F₃NO₂
Peso molecolare: 183,13 g/mol
MDL Number: MFCD00068195
CAS Number: 372-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfb2ba4a08c43b326f8026550cfe5c9e4e27b264&vid=dfb2ba4a08c43b326f8026550cfe5c9e4e27b264&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Freezing point 22.0 to 26.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR