Stampa…

4-Nitrophenyl trifluoroacetate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 4-Nitrophenyl 2,2,2-trifluoroacetate, Trifluoroacetic acid 4-Nitrophenyl ester

T0681-25G T0681-5G
TCIAT0681-25GEA 65 EUR
TCIAT0681-25G TCIAT0681-5G
4-Nitrophenyl trifluoroacetate ≥98.0% (by GC)
4-Nitrophenyl trifluoroacetate
Formula: C₈H₄F₃NO₄
Peso molecolare: 235.1200 g/mol
Punto di ebollizione: 140°C
MDL Number: MFCD00007324
CAS Number: 658-78-6
EINECS: 211-524-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa5fabc12fd2edfd07e1fd81732d6bd5f80211e7&vid=aa5fabc12fd2edfd07e1fd81732d6bd5f80211e7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 35.0 to 39.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR