Stampa…

Diphenyl phosphite ≥75.0% (by HPLC) (contains ca. 5% Phenol)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Diphenoxyphosphine oxide, NSC 43786, Diphenyl phosphonate

D0907-25G D0907-500G D0907-100G
TCIAD0907-25GEA 18 EUR
TCIAD0907-25G TCIAD0907-500G TCIAD0907-100G
Diphenyl phosphite ≥75.0% (by HPLC) (contains ca. 5% Phenol)
Diphenyl phosphite
Formula: C₁₂H₁₁O₃P
Peso molecolare: 234,19 g/mol
Punto di ebollizione: 148°C
Punto di fusione: 12°C
Flash Pt: 186°C
MDL Number: MFCD00044497
CAS Number: 4712-55-4
EINECS: 225-202-8
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f48535dff456396074a2370dd67837203e6614a&vid=0f48535dff456396074a2370dd67837203e6614a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (HPLC) min. 75.0 area%
Phenol 3.0 and le 6.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR