Stampa…

(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine ≥97.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: (S)-N,N-Dimethyl-1-ferrocenylethylamine

D2529-1G D2529-200MG
TCIAD2529-1GEA 165 EUR
TCIAD2529-1G TCIAD2529-200MG
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine ≥97.0% (by GC)
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine
Formula: C₁₄H₁₉FeN
Peso molecolare: 257.16 g/mol
Flash Pt: 95°C
MDL Number: MFCD00066272
CAS Number: 31886-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=305a095b983e250693a236573faf949fd3586ae0&vid=305a095b983e250693a236573faf949fd3586ae0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to Amber to Dark purple clear liquid to cloudy liquid
Purity (GC) min. 97.0 %
Optical purity (LC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR