Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Diethyl [(ethoxycarbonyl)(fluoro)methyl]phosphonate, Triethyl 2-fluoro-2-phosphonoacetate
F0340-1G
F0340-5G
TCIAF0340-1GEA
28
EUR
TCIAF0340-1G
TCIAF0340-5G
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) ≥95.0% (by GC)
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent)
Formula:
C₈H₁₆FO₅P Peso molecolare: 242,19 g/mol Punto di ebollizione: 200°C Flash Pt: 73°C |
MDL Number:
MFCD00134400 CAS Number: 2356-16-3 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...