Diethyl (2-oxopropyl)phosphonate ≥95.0% (by GC)
Fornitore: TCI
Synonyms:
Diethyl acetylmethylphosphonate
Formula:
C₇H₁₅O₄P Punto di ebollizione: 126°C |
MDL Number:
MFCD00044728 CAS Number: 1067-71-6 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...