4-Chloro-6-(trifluoromethyl)quinoline ≥99%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 99%
CAS No.: 49713-56-6
Molecular Formula: C10H5ClF3N
Formula Weight: 231.61
MDL No.: MFCD00153106
Formula:
C₁₀H₅ClF₃N Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00153106 CAS Number: 49713-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...