Stampa…

n-Butyl phenyl ether ≥99.0% (by GC)

Fornitore: TCI

Synonyms: N-butylphenylether

B0927-25ML B0927-500ML
TCIAB0927-25MLEA 40 EUR
TCIAB0927-25ML TCIAB0927-500ML
n-Butyl phenyl ether ≥99.0% (by GC)
n-Butyl phenyl ether
Formula: C₁₀H₁₄O
Punto di ebollizione: 210°C
Flash Pt: 82°C
MDL Number: MFCD00009438
CAS Number: 1126-79-0
EINECS: 214-426-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb7942fbb070584ab3aa4c39286229b60259329a&vid=eb7942fbb070584ab3aa4c39286229b60259329a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR