o-Tolyl acetate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Acetic acid o-Tolyl ester, Acetic acid o-Cresyl ester, o-Cresyl acetate
A0669-25G
A0669-500G
TCIAA0669-25GEA
34
EUR
TCIAA0669-25G
TCIAA0669-500G
o-Tolyl acetate ≥98.0% (by GC)
o-Tolyl acetate
Formula:
C₉H₁₀O₂ Peso molecolare: 150,1700 g/mol |
MDL Number:
MFCD00053704 CAS Number: 533-18-6 EINECS: 208-556-8 Merck Index: 14,02584 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...