6-Amino-2-naphthoic acid ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A1938-25G
A1938-5G
TCIAA1938-25GEA
526
EUR
TCIAA1938-25G
TCIAA1938-5G
6-Amino-2-naphthoic acid ≥97.0% (by HPLC, titration analysis)
6-Amino-2-naphthoic acid
Formula:
C₁₁H₉NO₂ Peso molecolare: 187,1947 g/mol Punto di fusione: 225°C |
MDL Number:
MFCD01861831 CAS Number: 116668-47-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...