Stampa…

N-Ethylcyclohexylamine ≥99.0% (by GC)

Fornitore: TCI
Pericolo

E0506-25ML E0506-500ML
TCIAE0506-25MLEA 16 EUR
TCIAE0506-25ML TCIAE0506-500ML
N-Ethylcyclohexylamine ≥99.0% (by GC)
N-Ethylcyclohexylamine
Formula: C₈H₁₇N
Punto di ebollizione: 164°C
Flash Pt: 30°C
MDL Number: MFCD00003834
CAS Number: 5459-93-8
EINECS: 226-733-8
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53942c82aeb99a1ee3a1827464602684873dae03&vid=53942c82aeb99a1ee3a1827464602684873dae03&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR