Stampa…

(S)-(-)-4-Amino-2-hydroxybutyric acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

A1438-25G A1438-5G
TCIAA1438-25GEA 88 EUR
TCIAA1438-25G TCIAA1438-5G
(S)-(-)-4-Amino-2-hydroxybutyric acid ≥98.0% (by titrimetric analysis)
(S)-(-)-4-Amino-2-hydroxybutyric acid
Formula: C₄H₉NO₃
MDL Number: MFCD00674110
CAS Number: 40371-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6285981067e79088add86de962de78c0e284e197&vid=6285981067e79088add86de962de78c0e284e197&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -28.0 to -32.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR