Stampa…

6-Hydroxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 6-Hydroxy-1-naphthalenecarboxylic acid

H0886-25G H0886-5G
TCIAH0886-25GEA 643 EUR
TCIAH0886-25G TCIAH0886-5G
6-Hydroxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)
6-Hydroxy-1-naphthoic acid
Formula: C₁₁H₈O₃
Peso molecolare: 188.1800 g/mol
MDL Number: MFCD00209983
CAS Number: 2437-17-4
EINECS: 407-390-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adcc40b08be63dcbda0c6049ead27a1a5c3ccb45&vid=adcc40b08be63dcbda0c6049ead27a1a5c3ccb45&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 207.0 to 212.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR