Methyl-6-hydroxy-2-naphthoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
6-Hydroxy-2-naphthoic acid methyll ester, Methyl 6-hydroxy-2-naphthoate
H1053-25G
H1053-5G
TCIAH1053-25GEA
234
EUR
TCIAH1053-25G
TCIAH1053-5G
Methyl-6-hydroxy-2-naphthoate ≥98.0% (by GC)
Methyl-6-hydroxy-2-naphthoate
Formula:
C₁₂H₁₀O₃ Peso molecolare: 202,2060 g/mol |
MDL Number:
MFCD00100526 CAS Number: 17295-11-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...