Stampa…

tert-Butylferrocene ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: t-Butylferrocene

B2063-1G B2063-5G
TCIAB2063-1GEA 65 EUR
TCIAB2063-1G TCIAB2063-5G
tert-Butylferrocene ≥97.0% (by GC)
tert-Butylferrocene
Formula: C₁₄H₁₈Fe
Peso molecolare: 242,14 g/mol
MDL Number: MFCD00058729
CAS Number: 1316-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b5882a0dbcfd41a93800f4db693c23589d1f3d8&vid=8b5882a0dbcfd41a93800f4db693c23589d1f3d8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green clear liquid to cloudy liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR