Stampa…

Ferrocenecarbaldehyde ≥97%

Fornitore: Thermo Scientific

Synonyms: Ferrocenecarboxaldehyde

39254.04 39254.09
39254.04EA 72.3 EUR
39254.04 39254.09
Ferrocenecarbaldehyde ≥97%
Ferrocenecarbaldehyde

Purity: 97% CAS No.: 12093-10-6 Molecular Formula: C11H10FeO Formula Weight: 214.05 Physical Form: Crystalline Sensitivity: Air Sensitive MDL No.: MFCD00001429

Formula: C₁₁H₁₀FeO
Peso molecolare: 214,04 g/mol
Punto di fusione: 124 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00001429
CAS Number: 12093-10-6
EINECS: 235-158-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR