2-Coumaranone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Benzofuran-2(3H)-one, Benzo[b]furan-2(3H)-one
C1445-25G
C1445-5G
TCIAC1445-25GEA
45
EUR
TCIAC1445-25G
TCIAC1445-5G
2-Coumaranone ≥98.0% (by GC)
2-Coumaranone
Formula:
C₈H₆O₂ Peso molecolare: 134,13 g/mol Punto di ebollizione: 244°C |
MDL Number:
MFCD00005856 CAS Number: 553-86-6 EINECS: 209-052-0 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...