Diisobutyl phosphonate ≥95.0% (by GC)
Fornitore: TCI
|
Attenzione
|
D3417-25G
D3417-500G
TCIAD3417-25GEA
17
EUR
TCIAD3417-25G
TCIAD3417-500G
Diisobutyl phosphonate ≥95.0% (by GC)
Diisobutyl phosphonate
Formula:
C₈H₁₉O₃P Punto di ebollizione: 138°C |
CAS Number:
1189-24-8 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...