3-(Trifluoromethyl)phenylacetone ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
T1693-25G
T1693-5G
TCIAT1693-25GEA
53
EUR
TCIAT1693-25G
TCIAT1693-5G
3-(Trifluoromethyl)phenylacetone ≥98.0% (by GC)
3-(Trifluoromethyl)phenylacetone
Formula:
C₁₀H₉F₃O Flash Pt: 90°C |
MDL Number:
MFCD00000397 CAS Number: 21906-39-8 EINECS: 244-652-6 |
|
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...